siRNA Details
| Gene Symbol: Asic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034014.1 | NP_001029186.1 | 2 | 731 | ||
| NM_012892.2 | NP_037024.2 | 2 | 746 | |||
| GenBank mRNA | U53211.1 | - | 2 | 731 | ||
| Y14635.1 | - | 2 | 746 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asic2| Gene Symbol: Asic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034014.1 | NP_001029186.1 | 2 | 731 | ||
| NM_012892.2 | NP_037024.2 | 2 | 746 | |||
| GenBank mRNA | U53211.1 | - | 2 | 731 | ||
| Y14635.1 | - | 2 | 746 | |||