siRNA Details
| Gene Symbol: Actg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012893.1 | NP_037025.1 | 7 | 899 | ||
| XM_006236728.2 | XP_006236790.1 | 8 | 1020 | |||
| GenBank mRNA | BC087689.1 | - | 8 | 965 | 7310913 | |
| FQ220056.1 | - | 8 | 964 | |||
| FQ220823.1 | - | 8 | 965 | |||
| FQ221094.1 | - | 8 | 964 | |||
| FQ221892.1 | - | 8 | 964 | |||
| FQ223160.1 | - | 8 | 990 | |||
| FQ229904.1 | - | 8 | 964 | |||
| M22323.1 | - | 7 | 899 | |||

Actg2
Genomic Map