siRNA Details
Gene Symbol: Acvr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031554.1 | NP_113742.1 | 4 | 695 | ||
XM_006244072.2 | XP_006244134.1 | 4 | 725 | |||
GenBank mRNA | BC097358.1 | - | 4 | 695 | 7442221 | |
L10640.1 | - | 2 | 139 | |||
M87067.1 | - | 4 | 932 |
