siRNA Details
| Gene Symbol: Adk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012895.3 | NP_037027.2 | 5 | 334 | ||
| XM_006251631.1 | XP_006251693.1 | 5 | 448 | |||
| GenBank mRNA | BC081712.1 | - | 5 | 334 | 7193044 | |
| U57042.1 | - | 4 | 274 | |||
| U90340.1 | - | 5 | 337 | |||

Adk
Genomic Map