siRNA Details
| Gene Symbol: SLC5A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006933.6 | NP_008864.4 | 2 | 1160 | ||
| GenBank mRNA | AX750578.1 | - | 2 | 1113 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC5A3| Gene Symbol: SLC5A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006933.6 | NP_008864.4 | 2 | 1160 | ||
| GenBank mRNA | AX750578.1 | - | 2 | 1113 | ||