siRNA Details
| Gene Symbol: Adprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183325.2 | NP_899154.2 | 4 | 897 | ||
| XM_006248360.2 | XP_006248422.1 | 4 | 1113 | |||
| GenBank mRNA | BC082065.1 | - | 4 | 897 | 7132643 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adprh| Gene Symbol: Adprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183325.2 | NP_899154.2 | 4 | 897 | ||
| XM_006248360.2 | XP_006248422.1 | 4 | 1113 | |||
| GenBank mRNA | BC082065.1 | - | 4 | 897 | 7132643 | |