siRNA Details
| Gene Symbol: Ahsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012898.4 | NP_037030.1 | 1 | 219 | ||
| GenBank mRNA | M29758.1 | - | 1 | 219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ahsg| Gene Symbol: Ahsg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012898.4 | NP_037030.1 | 1 | 219 | ||
| GenBank mRNA | M29758.1 | - | 1 | 219 | ||