siRNA Details
| Gene Symbol: Aldh3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031972.1 | NP_114178.1 | 3 | 385 | ||
| GenBank mRNA | BC070924.1 | - | 3 | 418 | 7096634 | |
| J03637.1 | - | 3 | 385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aldh3a1| Gene Symbol: Aldh3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031972.1 | NP_114178.1 | 3 | 385 | ||
| GenBank mRNA | BC070924.1 | - | 3 | 418 | 7096634 | |
| J03637.1 | - | 3 | 385 | |||