siRNA Details
| Gene Symbol: Ambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012901.1 | NP_037033.1 | 4 | 396 | ||
| XM_008763759.1 | XP_008761981.1 | 4 | 482 | |||
| GenBank mRNA | BC088166.1 | - | 4 | 381 | 7368246 | |
| S87544.1 | - | 4 | 396 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ambp| Gene Symbol: Ambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012901.1 | NP_037033.1 | 4 | 396 | ||
| XM_008763759.1 | XP_008761981.1 | 4 | 482 | |||
| GenBank mRNA | BC088166.1 | - | 4 | 381 | 7368246 | |
| S87544.1 | - | 4 | 396 | |||