siRNA Details
| Gene Symbol: Anxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012904.2 | NP_037036.1 | 12 | 976 | ||
| XM_006231159.1 | XP_006231221.1 | 13 | 1144 | |||
| XM_008760295.1 | XP_008758517.1 | 12 | 1063 | |||
| GenBank mRNA | BC061710.1 | - | 12 | 974 | 5598112 | |
| FQ213489.1 | - | 12 | 991 | |||
| FQ215033.1 | - | 12 | 991 | |||
| FQ216134.1 | - | 12 | 991 | |||
| FQ228716.1 | - | 12 | 991 | |||
| FQ229071.1 | - | 12 | 991 | |||
| FQ229198.1 | - | 11 | 912 | |||
| M19967.1 | - | 11 | 943 | |||
| Y00446.1 | - | 12 | 977 | |||

Anxa1
Genomic Map