siRNA Details
| Gene Symbol: Anxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012904.2 | NP_037036.1 | 9 | 755 | ||
| XM_006231159.1 | XP_006231221.1 | 10 | 923 | |||
| XM_008760295.1 | XP_008758517.1 | 9 | 842 | |||
| GenBank mRNA | BC061710.1 | - | 9 | 753 | 5598112 | |
| FQ213489.1 | - | 9 | 770 | |||
| FQ215033.1 | - | 9 | 770 | |||
| FQ216134.1 | - | 9 | 770 | |||
| M19967.1 | - | 8 | 722 | |||
| Y00446.1 | - | 9 | 756 | |||

Anxa1
Genomic Map