siRNA Details
| Gene Symbol: Aqp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012909.2 | NP_037041.2 | 2, 3 | 607 | ||
| GenBank mRNA | BC128705.1 | - | 2, 3 | 593 | 7109642 | |
| D13906.1 | - | 2, 3 | 607 | |||
| L28112.1 | - | 2, 3 | 646 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aqp2| Gene Symbol: Aqp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012909.2 | NP_037041.2 | 2, 3 | 607 | ||
| GenBank mRNA | BC128705.1 | - | 2, 3 | 593 | 7109642 | |
| D13906.1 | - | 2, 3 | 607 | |||
| L28112.1 | - | 2, 3 | 646 | |||