siRNA Details
| Gene Symbol: Atp1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012913.1 | NP_037045.1 | 1 | 196 | ||
| GenBank mRNA | BC061719.1 | - | 1 | 199 | 5598284 | |
| D84450.1 | - | 1 | 196 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp1b3| Gene Symbol: Atp1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012913.1 | NP_037045.1 | 1 | 196 | ||
| GenBank mRNA | BC061719.1 | - | 1 | 199 | 5598284 | |
| D84450.1 | - | 1 | 196 | |||