siRNA Details
| Gene Symbol: SLC5A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014227.2 | NP_055042.1 | 14 | 1719 | ||
| XM_006724308.3 | XP_006724371.1 | 15 | 1970 | |||
| XM_011530342.2 | XP_011528644.1 | 16 | 2088 | |||
| XM_011530343.2 | XP_011528645.1 | 14 | 1634 | |||
| XM_011530344.2 | XP_011528646.1 | 13 | 1599 | |||
| XM_017028920.1 | XP_016884409.1 | 16 | 2049 | |||
| GenBank mRNA | AJ133127.1 | - | 14 | 1733 | ||
| GQ292716.1 | - | 3 | 447 | |||
| JQ340171.1 | - | 3 | 447 | |||
| JQ519903.1 | - | 3 | 447 | |||

SLC5A4
Genomic Map