siRNA Details
| Gene Symbol: Atpif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012915.2 | NP_037047.2 | 2 | 202 | ||
| GenBank mRNA | BC126065.1 | - | 2 | 167 | 7442425 | |
| D13122.1 | - | 2 | 202 | |||
| FQ221416.1 | - | 2 | 187 | |||
| FQ229412.1 | - | 2 | 164 | |||

Atpif1
Genomic Map