siRNA Details
| Gene Symbol: Camk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012920.1 | NP_037052.1 | 16 | 1179 | ||
| GenBank mRNA | J02942.1 | - | 16 | 1179 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Camk2a| Gene Symbol: Camk2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012920.1 | NP_037052.1 | 16 | 1179 | ||
| GenBank mRNA | J02942.1 | - | 16 | 1179 | ||