siRNA Details
| Gene Symbol: Ccng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012923.2 | NP_037055.1 | 6 | 1013 | ||
| GenBank mRNA | BC081852.1 | - | 6 | 984 | 7105829 | |
| FQ211577.1 | - | 5 | 986 | |||
| FQ211975.1 | - | 6 | 1065 | |||
| FQ224502.1 | - | 6 | 974 | |||
| X70871.1 | - | 6 | 983 | |||

Ccng1
Genomic Map