siRNA Details
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012924.2 | NP_037056.2 | 14, 15 | 1839 | ||
GenBank mRNA | U96138.1 | - | 14, 15 | 1839 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012924.2 | NP_037056.2 | 14, 15 | 1839 | ||
GenBank mRNA | U96138.1 | - | 14, 15 | 1839 |