siRNA Details
| Gene Symbol: Cnga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012928.1 | NP_037060.1 | 1711 | |||
| GenBank mRNA | AF126808.1 | - | 1605 | |||
| X55519.1 | - | 1711 | ||||
| Gene Symbol: LOC103690077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008759081.1 | XP_008757303.1 | 3 | 1063 | ||

Cnga2
Genomic Map