siRNA Details
Gene Symbol: Bcar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012931.1 | NP_037063.1 | 5 | 2321 | ||
XM_006255628.2 | XP_006255690.1 | 5 | 2165 | |||
XM_006255629.2 | XP_006255691.1 | 5 | 4122 | |||
GenBank mRNA | D29766.1 | - | 5 | 2321 | ||
FQ212404.1 | - | 5 | 2123 | |||
FQ221713.1 | - | 5 | 2109 |
