siRNA Details
| Gene Symbol: Crmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012932.2 | NP_037064.1 | 6, 7 | 835 | ||
| XM_006251084.2 | XP_006251146.1 | 6, 7 | 1069 | |||
| GenBank mRNA | U52095.1 | - | 6, 7 | 629 | ||
| U52102.1 | - | 6, 7 | 803 | |||

Crmp1
Genomic Map