siRNA Details
| Gene Symbol: Dpysl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105717.2 | NP_001099187.1 | 11 | 1402 | ||
| XM_006252097.2 | XP_006252159.1 | 11 | 1633 | |||
| GenBank mRNA | CA510768.1 | - | 5 | 519 | ||
| CB577285.1 | - | 2 | 150 | |||
| Z46882.1 | - | 11 | 1355 | |||

Dpysl2
Genomic Map