siRNA Details
| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012934.1 | NP_037066.1 | 12 | 1513 | ||
| XM_006254670.2 | XP_006254732.1 | 12 | 2093 | |||
| XM_006254671.2 | XP_006254733.1 | 12 | 1724 | |||
| GenBank mRNA | AF389425.1 | - | 12 | 1513 | ||
| AF398465.1 | - | 12 | 1782 | |||

Dpysl3
Genomic Map