siRNA Details
Gene Symbol: Crybb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012937.1 | NP_037069.1 | 4 | 403 | ||
GenBank mRNA | X16072.1 | - | 4 | 403 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Crybb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012937.1 | NP_037069.1 | 4 | 403 | ||
GenBank mRNA | X16072.1 | - | 4 | 403 |