siRNA Details
| Gene Symbol: Ctse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012938.1 | NP_037070.1 | 3, 4 | 430 | ||
| XM_006249748.2 | XP_006249810.1 | 3, 4 | 635 | |||
| XM_006249749.2 | XP_006249811.1 | 4, 5 | 693 | |||
| GenBank mRNA | BC062002.1 | - | 3, 4 | 440 | 5599861 | |
| D38104.1 | - | 3, 4 | 430 | |||
| D45187.1 | - | 3, 4 | 572 | |||

Ctse
Genomic Map