siRNA Details
| Gene Symbol: Dlx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012943.1 | NP_037075.1 | 1 | 398 | ||
| GenBank mRNA | AB073716.1 | - | 1 | 120 | ||
| D31734.1 | - | 1 | 395 | |||
| L24443.1 | - | 1 | 398 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlx5| Gene Symbol: Dlx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012943.1 | NP_037075.1 | 1 | 398 | ||
| GenBank mRNA | AB073716.1 | - | 1 | 120 | ||
| D31734.1 | - | 1 | 395 | |||
| L24443.1 | - | 1 | 398 | |||