siRNA Details
| Gene Symbol: Sparcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012946.1 | NP_037078.1 | 10 | 2185 | ||
| GenBank mRNA | BC061755.1 | - | 10 | 2186 | 5600008 | |
| U27562.1 | - | 10 | 2185 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sparcl1| Gene Symbol: Sparcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012946.1 | NP_037078.1 | 10 | 2185 | ||
| GenBank mRNA | BC061755.1 | - | 10 | 2186 | 5600008 | |
| U27562.1 | - | 10 | 2185 | |||