siRNA Details
| Gene Symbol: Fgf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012951.1 | NP_037083.1 | 3 | 474 | ||
| GenBank mRNA | D79215.1 | - | 3 | 474 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf10| Gene Symbol: Fgf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012951.1 | NP_037083.1 | 3 | 474 | ||
| GenBank mRNA | D79215.1 | - | 3 | 474 | ||