siRNA Details
| Gene Symbol: Fshb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007597.1 | NP_001007598.1 | 2 | 231 | ||
| XM_006234672.2 | XP_006234734.1 | 3 | 1306 | |||
| GenBank mRNA | BC168724.1 | - | 3 | 293 | 6889701 | |
| M36804.1 | - | 2 | 244 | |||

Fshb
Genomic Map