siRNA Details
| Gene Symbol: Fshr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199237.1 | NP_954707.1 | 8 | 681 | ||
| XM_008764476.1 | XP_008762698.1 | 6 | 341 | |||
| GenBank mRNA | L02842.1 | - | 8 | 681 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fshr| Gene Symbol: Fshr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199237.1 | NP_954707.1 | 8 | 681 | ||
| XM_008764476.1 | XP_008762698.1 | 6 | 341 | |||
| GenBank mRNA | L02842.1 | - | 8 | 681 | ||