siRNA Details
| Gene Symbol: Gabrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012956.1 | NP_037088.1 | 7 | 846 | ||
| XM_008770136.1 | XP_008768358.1 | 3 | 784 | |||
| GenBank mRNA | X15466.1 | - | 7 | 846 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrb1| Gene Symbol: Gabrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012956.1 | NP_037088.1 | 7 | 846 | ||
| XM_008770136.1 | XP_008768358.1 | 3 | 784 | |||
| GenBank mRNA | X15466.1 | - | 7 | 846 | ||