siRNA Details
| Gene Symbol: Gfra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012959.1 | NP_037091.1 | 3, 4 | 671 | ||
| XM_008760512.1 | XP_008758734.1 | 4, 5 | 814 | |||
| XM_008760513.1 | XP_008758735.1 | 4, 5 | 678 | |||
| GenBank mRNA | U59486.1 | - | 3, 4 | 716 | ||
| U97142.1 | - | 3, 4 | 671 | |||

Gfra1
Genomic Map