siRNA Details
| Gene Symbol: Gfra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012959.1 | NP_037091.1 | 7, 8 | 1263 | ||
| XM_008760512.1 | XP_008758734.1 | 8, 9 | 1406 | |||
| XM_008760513.1 | XP_008758735.1 | 8, 9 | 1270 | |||
| XM_008760514.1 | XP_008758736.1 | 6, 7 | 1286 | |||
| GenBank mRNA | AJ002072.1 | - | 6, 7 | 992 | ||
| U59486.1 | - | 7, 8 | 1308 | |||
| U97142.1 | - | 7, 8 | 1263 | |||

Gfra1
Genomic Map