siRNA Details
| Gene Symbol: Gpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012961.1 | NP_037093.1 | 1 | 55 | ||
| XM_008767077.1 | XP_008765299.1 | 2 | 249 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr1| Gene Symbol: Gpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012961.1 | NP_037093.1 | 1 | 55 | ||
| XM_008767077.1 | XP_008765299.1 | 2 | 249 | |||
Set Membership: |
|