siRNA Details
| Gene Symbol: Icam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012967.1 | NP_037099.1 | 5 | 1129 | ||
| GenBank mRNA | BC081837.1 | - | 5 | 1141 | 7122397 | |
| D00913.1 | - | 5 | 1129 | |||
| FQ228241.1 | - | 5 | 1149 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Icam1| Gene Symbol: Icam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012967.1 | NP_037099.1 | 5 | 1129 | ||
| GenBank mRNA | BC081837.1 | - | 5 | 1141 | 7122397 | |
| D00913.1 | - | 5 | 1129 | |||
| FQ228241.1 | - | 5 | 1149 | |||