siRNA Details
| Gene Symbol: Kcna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012970.3 | NP_037102.1 | 3 | 1944 | ||
| XM_006233132.2 | XP_006233194.1 | 3 | 2530 | |||
| XM_006233133.2 | XP_006233195.1 | 3 | 2112 | |||
| XM_006233134.2 | XP_006233196.1 | 3 | 2095 | |||
| XM_006233135.2 | XP_006233197.1 | 3 | 1899 | |||
| XM_008761371.1 | XP_008759593.1 | 3 | 2140 | |||
| GenBank mRNA | J04731.1 | - | 3 | 1944 | ||
| M74449.1 | - | 1 | 1413 | |||
| X16003.1 | - | 3 | 1925 | |||

Kcna2
Genomic Map