siRNA Details
| Gene Symbol: Lgals4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012975.1 | NP_037107.1 | 4 | 372 | ||
| XM_008759113.1 | XP_008757335.1 | 4 | 539 | |||
| GenBank mRNA | M73553.1 | - | 4 | 372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgals4| Gene Symbol: Lgals4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012975.1 | NP_037107.1 | 4 | 372 | ||
| XM_008759113.1 | XP_008757335.1 | 4 | 539 | |||
| GenBank mRNA | M73553.1 | - | 4 | 372 | ||