siRNA Details
| Gene Symbol: Lgals9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012977.1 | NP_037109.1 | 2, 3 | 219 | ||
| GenBank mRNA | BC061566.1 | - | 2, 3 | 180 | 6921383 | |
| U59462.1 | - | 2, 3 | 141 | |||
| U67958.1 | - | 2, 3 | 219 | |||
| U72741.1 | - | 2, 3 | 141 | |||

Lgals9
Genomic Map