siRNA Details
| Gene Symbol: Lhcgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012978.1 | NP_037110.1 | 5 | 473 | ||
| GenBank mRNA | M26199.1 | - | 5 | 473 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lhcgr| Gene Symbol: Lhcgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012978.1 | NP_037110.1 | 5 | 473 | ||
| GenBank mRNA | M26199.1 | - | 5 | 473 | ||