siRNA Details
| Gene Symbol: Man2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012979.2 | NP_037111.2 | 3 | 1441 | ||
| GenBank mRNA | FQ225400.1 | - | 3 | 621 | ||
| FQ225693.1 | - | 3 | 621 | |||
| M24353.1 | - | 3 | 358 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Man2a1| Gene Symbol: Man2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012979.2 | NP_037111.2 | 3 | 1441 | ||
| GenBank mRNA | FQ225400.1 | - | 3 | 621 | ||
| FQ225693.1 | - | 3 | 621 | |||
| M24353.1 | - | 3 | 358 | |||