siRNA Details
| Gene Symbol: Nfia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012988.2 | NP_037120.1 | 1 | 498 | ||
| GenBank mRNA | AB060652.1 | - | 406 | |||
| AF112455.1 | - | 578 | ||||
| AF112456.1 | - | 1 | 291 | |||
| AY572794.1 | - | 1 | 406 | |||
| D78017.1 | - | 1 | 498 | |||
| D78018.1 | - | 1 | 493 | |||
| D78019.1 | - | 1 | 478 | |||
| X13167.1 | - | 1 | 340 | |||

Nfia
Genomic Map