siRNA Details
| Gene Symbol: Nog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012990.1 | NP_037122.1 | 1 | 1186 | ||
| GenBank mRNA | BF549968.1 | - | 1 | 207 | ||
| U31203.1 | - | 1 | 673 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nog| Gene Symbol: Nog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012990.1 | NP_037122.1 | 1 | 1186 | ||
| GenBank mRNA | BF549968.1 | - | 1 | 207 | ||
| U31203.1 | - | 1 | 673 | |||