siRNA Details
| Gene Symbol: Npm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012992.4 | NP_037124.1 | 2 | 219 | ||
| XM_008767546.1 | XP_008765768.1 | 2 | 332 | |||
| GenBank mRNA | BC060579.1 | - | 2 | 223 | 6888417 | |
| BC088088.1 | - | 2 | 209 | 7382200 | ||
| J03969.1 | - | 2 | 172 | |||
| J04943.1 | - | 2 | 201 | |||

Npm1
Genomic Map