siRNA Details
| Gene Symbol: Nrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012993.2 | NP_037125.1 | 18 | 2169 | ||
| XM_006238499.2 | XP_006238561.1 | 20 | 2507 | |||
| XM_006238500.2 | XP_006238562.1 | 19 | 2371 | |||
| GenBank mRNA | FQ218095.1 | - | 18 | 2292 | ||
| L27124.1 | - | 18 | 2170 | |||
| X93208.1 | - | 20 | 2374 | |||

Nrd1
Genomic Map