siRNA Details
| Gene Symbol: P4hb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012998.1 | NP_037130.1 | 8, 9 | 1216 | ||
| GenBank mRNA | BC061857.1 | - | 8, 9 | 1230 | 5623688 | |
| FQ209800.1 | - | 1, 2 | 30 | |||
| M21018.1 | - | 8, 9 | 1094 | |||
| X02918.1 | - | 8, 9 | 1216 | |||

P4hb
Genomic Map