siRNA Details
| Gene Symbol: Pam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013000.2 | NP_037132.2 | 2 | 307 | ||
| XM_006245594.2 | XP_006245656.1 | 2 | 520 | |||
| XM_006245595.2 | XP_006245657.1 | 2 | 520 | |||
| XM_006245596.2 | XP_006245658.1 | 2 | 520 | |||
| XM_006245597.2 | XP_006245659.1 | 2 | 520 | |||
| XM_006245598.2 | XP_006245660.1 | 2 | 519 | |||
| XM_006245599.2 | XP_006245661.1 | 2 | 519 | |||
| XM_008767360.1 | XP_008765582.1 | 2 | 520 | |||
| GenBank mRNA | M25719.1 | - | 2 | 423 | ||
| M25732.1 | - | 2 | 307 | |||
| M82845.1 | - | 2 | 178 | |||
| X59689.1 | - | 2 | 128 | |||

Pam
Genomic Map