siRNA Details
| Gene Symbol: Pax6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013001.2 | NP_037133.1 | 7 | 798 | ||
| XM_006234633.2 | XP_006234695.1 | 9 | 1223 | |||
| XM_006234634.2 | XP_006234696.1 | 9 | 2679 | |||
| XM_006234635.2 | XP_006234697.1 | 9 | 1138 | |||
| XM_006234636.2 | XP_006234698.1 | 9 | 2129 | |||
| XM_008762060.1 | XP_008760282.1 | 8 | 1322 | |||
| GenBank mRNA | AY540905.1 | - | 6 | 695 | ||
| AY540906.1 | - | 6 | 695 | |||
| BC128741.1 | - | 7 | 798 | 8363963 | ||
| HQ121510.1 | - | 5 | 681 | |||
| U69644.1 | - | 7 | 1018 | |||

Pax6
Genomic Map