siRNA Details
| Gene Symbol: Pcp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC059147.1 | - | 2 | 108 | 6888437 | |
| BG665749.1 | - | 2 | 107 | |||
| BG666168.1 | - | 2 | 98 | |||
| CB613293.1 | - | 2 | 103 | |||
| M24852.1 | - | 2 | 95 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcp4| Gene Symbol: Pcp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC059147.1 | - | 2 | 108 | 6888437 | |
| BG665749.1 | - | 2 | 107 | |||
| BG666168.1 | - | 2 | 98 | |||
| CB613293.1 | - | 2 | 103 | |||
| M24852.1 | - | 2 | 95 | |||