siRNA Details
| Gene Symbol: Pik3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013005.1 | NP_037137.1 | 13 | 1793 | ||
| XM_006231868.2 | XP_006231930.1 | 8 | 1979 | |||
| XM_008760659.1 | XP_008758881.1 | 8 | 3429 | |||
| GenBank mRNA | D64045.1 | - | 13 | 1793 | ||
| D64048.1 | - | 8 | 983 | |||
| D78486.1 | - | 8 | 893 | |||
| U50412.1 | - | 8 | 987 | |||

Pik3r1
Genomic Map