siRNA Details
| Gene Symbol: Pnoc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013007.1 | NP_037139.1 | 2 | 419 | ||
| XM_006252196.2 | XP_006252258.1 | 3 | 581 | |||
| XM_006252199.2 | XP_006252261.1 | 3 | 579 | |||
| GenBank mRNA | FQ214340.1 | - | 3 | 466 | ||
| FQ220805.1 | - | 3 | 468 | |||
| S79730.1 | - | 2 | 356 | |||
| U48262.1 | - | 2 | 422 | |||
| X97375.1 | - | 2 | 419 | |||

Pnoc
Genomic Map